Skip to Content
Merck
All Photos(1)

Key Documents

337501

Sigma-Aldrich

[Tris(dimethylphenylphosphine)](2,5-norbornadiene)rhodium(I) hexafluorophosphate

97%

Synonym(s):

[(2,3-η)-Bicyclo[2.2.1]hepta-2,5-diene]tris(dimethylphenylphosphine)rhodium(I) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H41F6P4Rh
CAS Number:
Molecular Weight:
754.45
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: rhodium
reagent type: catalyst

mp

158-161 °C (lit.)

SMILES string

[Rh+].F[P-](F)(F)(F)(F)F.[CH]1[CH]C2[CH][CH]C1C2.CP(C)c3ccccc3.CP(C)c4ccccc4.CP(C)c5ccccc5

InChI

1S/3C8H11P.C7H8.F6P.Rh/c3*1-9(2)8-6-4-3-5-7-8;1-2-7-4-3-6(1)5-7;1-7(2,3,4,5)6;/h3*3-7H,1-2H3;1-4,6-7H,5H2;;/q;;;;-1;+1

InChI key

KSBHDOZWEREXLG-UHFFFAOYSA-N

Application

Catalyst for aminomethylation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaspard Huber et al.
Magnetic resonance in chemistry : MRC, 56(7), 672-678 (2017-12-09)
Cryptophanes, cage-molecules constituted of aromatic bowls, are now well recognised as powerful xenon hosts in 129 Xe NMR-based biosensing. In the quest of a dual probe that can be addressed only by NMR, we have studied three cryptophanes bearing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service