Skip to Content
Merck
All Photos(1)

Documents

275204

Sigma-Aldrich

3,3,3-Tris(4-chlorophenyl)propionitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)3CCH2CN
CAS Number:
Molecular Weight:
386.70
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

175-177 °C (lit.)

SMILES string

Clc1ccc(cc1)C(CC#N)(c2ccc(Cl)cc2)c3ccc(Cl)cc3

InChI

1S/C21H14Cl3N/c22-18-7-1-15(2-8-18)21(13-14-25,16-3-9-19(23)10-4-16)17-5-11-20(24)12-6-17/h1-12H,13H2

InChI key

SGXWKWVVMYKJNN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service