All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12162002
Recommended Products
Assay
97%
refractive index
n20/D 1.435 (lit.)
density
0.854 g/mL at 25 °C (lit.)
SMILES string
C=CCCCCC1CO1
Packaging
Packaged in glass bottles
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
100.4 °F - closed cup
Flash Point(C)
38 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service