Skip to Content
Merck
All Photos(1)

Key Documents

254053

Sigma-Aldrich

Praseodymium(III) nitrate hexahydrate

≥99.99% trace metals basis

Synonym(s):

Praseodymium trinitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr(NO3)3·6H2O
CAS Number:
Molecular Weight:
435.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: praseodymium

SMILES string

O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Pr/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

LXXCECZPOWZKLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Hydration may vary slightly

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Devaraj Ramasamy et al.
Physical chemistry chemical physics : PCCP, 17(17), 11527-11539 (2015-04-11)
The current work demonstrates how tailoring the transport properties of thin ceria-based buffer layers in solid oxide fuel or electrolyser cells can provide the necessary phase stability against chemical interaction at the electrolyte/electrode interface, while also providing radical improvements in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service