251666
1-(3-Sulfopropyl)-2-vinylpyridinium hydroxide inner salt
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H13NO3S
CAS Number:
Molecular Weight:
227.28
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Assay
97%
mp
220 °C (dec.) (lit.)
SMILES string
[O-]S(=O)(=O)CCC[n+]1ccccc1C=C
InChI
1S/C10H13NO3S/c1-2-10-6-3-4-7-11(10)8-5-9-15(12,13)14/h2-4,6-7H,1,5,8-9H2
InChI key
DNHDSWZXBHTLDP-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service