Skip to Content
Merck
All Photos(1)

Key Documents

132926

Sigma-Aldrich

Acetylsalicylic acid

99%

Synonym(s):

2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

mp

134-136 °C (lit.)

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels Hulsman et al.
Journal of medicinal chemistry, 50(10), 2424-2431 (2007-04-20)
Hybrid drug 1 (NO-ASA) continues to attract intense research from chemists and biologists alike. It consists of ASA and a -ONO2 group connected through a spacer and is in preclinical development as an antitumor drug. We report that, contrary to
Morshed Alam Chowdhury et al.
Bioorganic & medicinal chemistry letters, 18(23), 6138-6141 (2008-10-24)
A hitherto unknown class of celecoxib analogs was designed for evaluation as dual inhibitors of the 5-lipoxygenase/cyclooxygenase-2 (5-LOX/COX-2) enzymes. These compounds possess a SO(2)Me (11a), or SO(2)NH(2) (11b) COX-2 pharmacophore at the para-position of the N(1)-phenyl ring in conjunction with
Carlos A Velázquez et al.
Journal of medicinal chemistry, 51(6), 1954-1961 (2008-03-05)
The carboxylic acid group of the anti-inflammatory (AI) drugs aspirin and indomethacin was covalently linked to the 1-(2-carboxypyrrolidin-1-yl)diazen-1-ium-1,2-diolate ion via a one-carbon methylene spacer to obtain two new hybrid prodrugs. The aspirin prodrug ( 23) was a 2.2-fold more potent
Carlos Velázquez et al.
Journal of medicinal chemistry, 48(12), 4061-4067 (2005-06-10)
A novel group of hybrid nitric oxide-releasing nonsteroidal antiinflammatory drugs ((*)NO-NSAIDs) possessing a 1-(pyrrolidin-1-yl)diazen-1-ium-1,2-diolate (11, 13, 15) or 1-(N,N-dimethylamino)diazen-1-ium-1,2-diolate (12, 14, 16) moiety attached via a one-carbon methylene spacer to the carboxylic acid group of the traditional NSAIDs aspirin, ibuprofen
Morshed A Chowdhury et al.
Bioorganic & medicinal chemistry letters, 20(4), 1324-1329 (2010-01-26)
A new group of hybrid nitric oxide (NO) releasing anti-inflammatory (AI) coxib prodrugs (NO-coxibs) wherein the para-tolyl moiety present in celecoxib was replaced by a N-(4-nitrooxybutyl)piperidyl 15a-b, or N-(4-nitrooxybutyl)-1,2,3,6-tetrahydropyridyl 17a-b, NO-donor moiety was synthesized. All compounds released a low amount

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service