Skip to Content
Merck
All Photos(1)

Documents

116289

Sigma-Aldrich

4-Chloro-3-nitroanisole

98%

Synonym(s):

1-Chloro-4-methoxy-2-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)OCH3
CAS Number:
Molecular Weight:
187.58
Beilstein:
640872
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

41-43 °C (lit.)

SMILES string

COc1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C7H6ClNO3/c1-12-5-2-3-6(8)7(4-5)9(10)11/h2-4H,1H3

InChI key

HISHUMDTGXICEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-chloro-3-nitroanisole is used to synthesize a group of derivatives of 7-methanesulfonylamino-6-phenoxychromone at the pyrone and phenoxy rings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Inaba et al.
Chemical & pharmaceutical bulletin, 48(1), 131-139 (2000-03-08)
A group of derivatives of 7-methanesulfonylamino-6-phenoxychromone (1) at the pyrone and phenoxy rings was synthesized starting with 4-chloro-3-nitroanisole and evaluated against acute and chronic inflammations in oral administration in animals. Significant potency in the rat models of carrageenin-induced edema (CPE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service