MilliporeSigma
  • Kinetics of proinflammatory cytokines after intraperitoneal injection of tribromoethanol and a tribromoethanol/xylazine combination in ICR mice.
Kinetics of proinflammatory cytokines after intraperitoneal injection of tribromoethanol and a tribromoethanol/xylazine combination in ICR mice.
Laboratory animal research (2011-10-15)
Yoon Ju Cho, Young Ah Lee, Jae Won Lee, Jong In Kim, Jin Soo Han
ABSTRACT
Tribromoethanol (2,2,2-tribromoethanol, TBE) is a popular injectable anesthetic agent used in mice in Korea. Our goal was to assess the risks associated with side effects (lesions) in the abdominal cavity, especially at high doses. To understand the underlying pathophysiological changes, we examined levels of cytokines through ELISA of abdominal lavage fluid and spleen collected from mice treated with low and high-dose TBE. ICR mice were anesthetized using one of the following protocols: a combination of TBE 200 mg/kg (1.25%) and xylazine 10 mg/kg; TBE 400 mg/kg (1.25%); and TBE 400 mg/kg (2.5%). Administration of high-dose TBE (400 mg/kg) increased the interleukin-1β and interleukin-6 levels in the peritoneal cavity over the short term (<1 day) compared with sham controls and low-dose TBE (200 mg/kg) groups. Cytokine expression in the low-dose TBE group was similar to the control group, whereas in the high-dose TBE group cytokine levels were higher in abdominal lavage fluid and spleen over the long term (10 days post-injection). We conclude that a combination of TBE 200 mg/kg (1.25%) and xylazine (10 mg/kg) is a safe and effective anesthetic for use in animals.
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4