Merck
  • Ab initio calculations on low-lying electronic states of TeO2 and Franck-Condon simulation of the (1)1B2 <-- X1A1 TeO2 absorption spectrum including anharmonicity.
Ab initio calculations on low-lying electronic states of TeO2 and Franck-Condon simulation of the (1)1B2 <-- X1A1 TeO2 absorption spectrum including anharmonicity.
The Journal of chemical physics (2004-08-05)
Edmond P F Lee, Daniel K W Mok, Foo-tim Chau, John M Dyke
ABSTRACT
Ab initio calculations have been carried out on low-lying singlet and triplet states of TeO2 at different levels of theory with basis sets of up to the augmented-polarized valence-quintuple-zeta quality. Equilibrium geometrical parameters, harmonic vibrational frequencies, and relative electronic energies of the X1A1, 1B1, 1B2, 1A2, 3A1, 3B1, 3B2, and 3A2 states of TeO2 have been calculated. Potential energy functions (PEFs) of the X1A1 and the (1)1B2 states were computed at the complete-active-space self-consistent-field multireference configuration interaction level, with a basis set of augmented-polarized valence-quadruple-zeta quality. Franck-Condon factors (FCFs) for the electronic transition between the X1A1 and (1)1B2 states of TeO2 were calculated with the above-mentioned ab initio PEFs. The (1)1B2 <-- X1A1 absorption spectrum of TeO2 was simulated employing the computed FCFs, which include Duschinsky rotation and anharmonicity, and compared with the recently published laser-induced fluorescence (LIF) spectrum of Hullah and Brown [J. Mol. Spectrosc. 200, 261 (2000)]. The ab initio results and spectral simulation reported here confirm the upper electronic state involved in the LIF spectrum to be the (1)1B2 state of TeO2 and also confirm the vibrational assignments of Hullah and Brown. However, our simulated spectrum suggests that the reported LIF spectrum from 345 to 406 nm represents only a portion of the full (1)1B2 <-- X1A1 absorption spectrum of TeO2, which extends from ca. 406 to 300 nm. Another dye other than the two used by Hullah and Brown is required to cover the 345-300 nm region of the LIF band. Ab initio calculations show strong configuration mixing of the (1)1B2 electronic surface with higher 1B2 states in a region of large TeO bond length (> or = 2.0 A) and OTeO bond angle (> or = 135.0 degrees).
Sigma-Aldrich
Tellurium dioxide, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tellurium dioxide, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis