
Lithium bis(fluorosulfonyl)imide

Synonym(s):
LiFSI, Lithium bis(fluorosulfonyl)amide, Lithium bis(fluorosulfonyl)imido, Lithium imidodisulfuryl fluoride
Empirical Formula (Hill Notation):
F2LiNO4S2
CAS Number:
Molecular Weight:
187.07
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing