Skip to Content
Merck
All Photos(1)

Key Documents

20785-U

Hamilton® GASTIGHT® syringe, 1700 series, removable needle

1750RN, volume 0.5 mL, needle size 22 ga (bevel tip), needle L 51 mm (2 in.)

Synonym(s):

TM=["Hamilton"] Airtight syringe, 1700 series, detachable needle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500
NACRES:
NB.03
Pricing and availability is not currently available.

material

bevel tip stainless steel needle (point style 2)
glass body

needle size

22 ga (bevel tip)

feature

removable needle RN

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 81230

needle L

51 mm (2 in.)

volume

0.5 mL

compatibility

for use with Hamilton 1750RN

Looking for similar products? Visit Product Comparison Guide

Legal Information

GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Dear seller, could you please send me a photo of this syringe? I am looking for gas tight syringe ideally with lock on top. I used before which had red/green lock on top of the syringe. I would really appreciate your reply.

    1 answer
    1. See the product image below. This product does not include a push button stop valve, nor do any of the Hamilton® GASTIGHT® removable needle (RN) syringe models.

      A push button valve of this type is sold under Product No. 22285, but this valve is intended for use with the Hamilton® GASTIGHT® PTFE Luer Lock (TLL) syringes. The equivalent syringe with the TLL termination is sold under Product No. 20916. The needles compatible with this syringe and push button stop valve are sold under Product No. 21747 (KEL-F fluorocarbon polymer needle hub) or Product No. 21746 (metal needle hub).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service