11336045001
Roche
DNA Molecular Weight Marker VIII
pkg of 50 μg (in 200 μl), solution
Synonym(s):
DNA marker
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105335
form
solution
Quality Level
packaging
pkg of 50 μg (in 200 μl)
manufacturer/tradename
Roche
concentration
250 μg/mL
color
colorless
solubility
water: miscible
storage temp.
−20°C
Related Categories
General description
DNA Molecular Weight Marker VIII contains a mixture of 17 DNA fragments.
The product is also available as DNA Molecular Weight Marker VIII, Digoxigenin-labeled.
Size range: 19 to 1114 bp
The product is also available as DNA Molecular Weight Marker VIII, Digoxigenin-labeled.
Size range: 19 to 1114 bp
Application
DNA Molecular Weight Marker VIII is used for the size determination of DNA in agarose gels by:
Note: Use this marker to size DNA fragments between 19 and 1,114 bp.
This marker is also available in DIG-labeled form (DNA Molecular Weight Marker VIII, DIG-labeled).
- Restriction digests
- PCR
- RT-PCR
Note: Use this marker to size DNA fragments between 19 and 1,114 bp.
This marker is also available in DIG-labeled form (DNA Molecular Weight Marker VIII, DIG-labeled).
Features and Benefits
Fragment mixture prepared by cleavage of pUCBM21 DNA with Hpa II, and pUCBM21 DNA with Dra I and Hind III.
Contents
Ready-to-use solution, 250μg/ml, in TE buffer (10mM Tris-HCl, 1mM EDTA, pH 8.0).
Contents
Ready-to-use solution, 250μg/ml, in TE buffer (10mM Tris-HCl, 1mM EDTA, pH 8.0).
Sequence
The mixture contains 17 DNA fragments with the following base pair lengths (1 base pair = 660 daltons): 19, 26, 34/34, 37, 67, 110, 124, 147, 190, 242, 320, 404, 489, 501, 692, 900, and 1114 bp.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.
Unit Definition
50 μg = 1A260 unit
Preparation Note
Storage conditions (working solution): 2 to 8 °C
Once thawed we recommend further storage at 2 to 8 °C.
Once thawed we recommend further storage at 2 to 8 °C.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Use this marker to size DNA fragments between 19 and 1,114 bp.
This marker is also available in DIG-labeled form (DNA Molecular Weight Marker VIII, DIG-labeled).
This marker is also available in DIG-labeled form (DNA Molecular Weight Marker VIII, DIG-labeled).
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromosomal DNA damage in APOE ?4 carriers and noncarriers does not appear to be different.
Chua A, et al.
Environmental and Molecular Mutagenesis, 56(8), 694-708 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service