Skip to Content
Merck
All Photos(1)

Key Documents

MABF436

Sigma-Aldrich

Anti-CD45.2 Antibody (mouse), APC-Cy7, clone 104

clone 104, 0.2 mg/mL, from mouse

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

APC-Cy7

antibody form

purified antibody

antibody product type

primary antibodies

clone

104, monoclonal

species reactivity

mouse

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

General description

The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.

Application

This mouse monoclonal antibody Anti-CD45.2 (mouse), APC-Cy7, clone 104 validated for use in IP, Flow Cytometry & ICC for the detection of CD45.2.

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 μg of this antibody detected CD45.2 in one million C57Bl/6 splenocytes.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service