Skip to Content
Merck
All Photos(1)

Key Documents

ABN2264

Sigma-Aldrich

Anti-SNTF (cleaved spectrin)

serum, from rabbit

Synonym(s):

Spectrin alpha chain non-erythrocytic 1, Alpha-II spectrin, Fodrin alpha chain, Spectrin non-erythroid alpha subunit, Calpain-derived alpha-II spectrin N-terminal fragment

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Pricing and availability is not currently available. Contact Technical Service

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, porcine

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

General description

Spectrin alpha chain, non-erythrocytic 1 (UniProt: Q13813; also known as Alpha-II spectrin, Fodrin alpha chain, Spectrin, non-erythroid alpha subunit) is encoded by the SPTAN1 (also known as NEAS, SPTA2) gene (Gene ID: 6709) in human. Non-erythroid Spectrin plays a role in stabilizing membrane structures, maintaining cell shape and linking the cytoskeleton to plasma membranes or intracellular vesicles. Alpha II-spectrin is a structural protein abundant in neurons of the central nervous system and is cleaved into signature fragments by proteases involved in necrotic and apoptotic cell death. It is one of the primary targets cleaved by activated caspases during apoptosis. The full-length protein can be cleaved to yield an N-terminal product of ~150 kDa, and C-terminal products of ~120 and ~35 kDa. Alpha II- spectrin is involved in secretion, interacts with calmodulin in a calcium-dependent manner and is thus candidate for the calcium-dependent movement of the cytoskeleton at the membrane. It has two calcium-binding regions (aa 2336-2347 and 2379-2390). Its mu-calpain cleavage site is localized at Tyr-Gly (aa 1176-1177). Phosphorylation of Tyrosine 1176 is shown to reduce sensitivity to cleavage by calpain in vitro. Plasma levels of calpain-cleaved Alpha II-Spectrin, N-terminal fragment (SNTF) serve as a useful marker for mild to severe traumatic brain injury and SNTF immunostaining can also reveal previously undetected traumatic brain injury. Mutations in SPTAN1 gene are known to cause early infantile epileptic encephalopathy that is characterized by seizures associated with hypsarrhythmia, profound mental retardation with lack of visual attention and speech development. (Ref.: Siman, R., et al. (2013). Front. Neurol. 4; Article 190; Johnson VE., et al (2016). Acta Neuropathol. 131(1); 115-135).

Specificity

This rabbit polyclonal antibody specifically detects calpain-generated neoepitope in SNTF. It targets a neoepitope of 6 amino acids at the carboxyl-end of the stable calpain-derived alpha-spectrin.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to calpain-generated neoepitope in SNTF (CAQQEVY).


Application

Anti-SNTF (cleaved spectrin), Cat. No. ABN2264, is a highly specific rabbit polyclonal antibody that targets cleaved Spectrin alpha chain, non-erythrocytic 1 and has been tested for use in ELISA, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected SNTF (cleaved spectrin) in swine and human brain sections (Johnson, V.E., et. al. (2016). Acta Neuropathol. 131(1):115-35).

Western Blotting Analysis: A representative lot detected SNTF (cleaved spectrin) in Western Blotting applications (Roberts-Lewis, J.M., et. al. (1994). J Neurosci. 14(6):3934-44).

Enzyme Immunoassay Analysis (ELISA): A representative lot detected SNTF (cleaved spectrin) in detection antibody for human serum samples (Siman, R., et. al. (2013). Front Neurol. 4:190).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in mouse brain extract incubated with calcium chloride to activate calpain.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Spectrin alpha chain, non-erythrocytic 1 in Mouse Brain extract incubated with calcium chloride to activate Calpain.

Target description

~160 kDa observed. Calculated molecular weight of full length protein is 284.54 kDa Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aicha Saadane et al.
The American journal of pathology, 191(10), 1805-1821 (2021-07-03)
This study tested the hypothesis that diabetes promotes a greater than normal cytosolic calcium level in rod cells that activates a Ca2+-sensitive protease, calpain, resulting in oxidative stress and inflammation, two pathogenic factors of early diabetic retinopathy. Nondiabetic and 2-month

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service