574102
Sulindac Sulfide
A cell-permeable, active metabolite of Sulindac.
Synonym(s):
Sulindac Sulfide, (Z)-5-Fluoro-2-methyl-1-[ p-(methylthio)benzylidene]indene-3-acetic Acid
About This Item
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
yellow-orange
solubility
DMSO: 25 mg/mL
ethanol: 5 mg/mL
shipped in
ambient
storage temp.
10-30°C
SMILES string
Fc1cc2c(cc1)\C(=C\c3ccc(cc3)SC)\C(=C2CC(=O)O)C
InChI
1S/C20H17FO2S/c1-12-17(9-13-3-6-15(24-2)7-4-13)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9+
InChI key
LFWHFZJPXXOYNR-RQZCQDPDSA-N
General description
Biochem/physiol Actions
Warning
Other Notes
Lim, J.T., et al. 1999. Biochem. Pharmacol. 58, 1097.
Herrmann, C., et al. 1998. Oncogene 17, 1769.
Picariello, L., et al. 1998. Eur. J. Pharmacol. 360, 105.
Arber, N., et al. 1997. Gastroenterology 113, 1892.
Meade, E.A., et al. 1993. J. Biol. Chem. 268, 6610.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service