17-10506
AccuChIP Acetyl-Histone H3 (Lys9) Chromatin Immunoprecipitation Internal Control
target-specific spike-in controls that make ChIP experiments more quantitative and accurate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.75
Recommended Products
Quality Level
manufacturer/tradename
Upstate®
technique(s)
ChIP: suitable (ChIP-seq)
immunoprecipitation (IP): suitable
shipped in
dry ice
Related Categories
Application
- Normalization of ChIP signals
- Normalization of ChIP-seq signals
- Analysis of antibody specificity
Components
AccuChIP H3K9Ac Spike-In Reagent (50 pg/uL )
AccuChIP H3K9Ac Detection Primer Mix (5 μM)
AccuChIP Dilution Buffer
AccuChIP H3K9Ac Detection Primer Mix (5 μM)
AccuChIP Dilution Buffer
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service