GE80-1106-55
Electrode holder for MultiPhor™ II electrophoresis unit
pack of 1 ea
About This Item
Recommended Products
grade
electrophoresis grade
packaging
pack of 1 ea
manufacturer/tradename
Cytiva 80110655
Related Categories
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service