CLS3383
Corning® HTS Transwell® 96 well permeable support system, accessory plates
96 well reservoir plate, 1-well reservoir, non-treated, flat bottom, sterile, w/lid, clear, 10/cs
Synonym(s):
corning transwell, transwell corning
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
clear polystyrene plate
flat bottom wells clear
polystyrene
rectangular wells
sterility
sterile
feature
lid
skirt
packaging
case of 10
pack of 10
manufacturer/tradename
Corning 3383
size
96 wells
well working volume
20-25 mL
color
clear
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Product is an accessory for the Corning® HTS Transwell®-96 Permeable Support system
- Open plate for cell growth and feeding stage, sterile, not treated (for cell attachment)
- Removable sterile media stabilizer included to minimize sloshing during movement of plates
- Used to minimize contamination when change of media is needed or to prevent cross contamination when media type is switched during cell growth step
- When used with HTS Transwell®-96 unit, the tray allows for convenient multi-channel pipette access (manual or automated) along the A1- A8 side
- SBS standard footprint
- Sterile lid included with each tray
Legal Information
Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service