Skip to Content
Merck
All Photos(1)

Key Documents

TPS

96112839, fish kidney (anterior), Fibroblast-like/Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

TPS, 96112839

biological source

fish kidney (anterior)

growth mode

Adherent

karyotype

Modal no. 58

morphology

Fibroblast-like/Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish Rainbow Trout Pronephric Stroma

Cell Line Description

TPS has been initiated from a long-term pronephric stroma culture of an adult 1 year old rainbow trout (Oncorhynchus mykiss W). The cells grow at 4° to 22°C, with an optimum temperature of 18°C. They do not form true monolayers, but overlap in a multilayered network of fibroblastic, epithelioid and giant cells. TPS cells are susceptible to viral haemorrhagic septicemia virus (VHSV) and infectious haemopoietic necrosis virus (IHNV). They have been applied to support the development of haemopoiesis when co-cultured with haemopoietic precursors. Spontaneous angiogenesis processes in vitro occurred in TPS cultures when they are kept in the same flask, i.e. without passages, for long times (over 2 months).

Application

Fish Haemopoiesis and Fish pathology.

Culture Medium

RPMI 1640 + 2mM Glutamine + 25mM Hepes + 2.5mM NaP + 0.01mM Hydrocortisone-21-hemisuccinate + 0.088mM Guanosine + 0.0225mM Cytidine + 0.094mM Adenosine + 0.102mM Uridine + 10% FBS + 10% HS, pH 7.4-7.6. At ECACC successful growth of this cell line has bee

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 using 0.25% trypsin or trypsin/EDTA; trypsinise at room temperature once a week; air atmosphere; 18°C. Fish cell lines detach easily during transit if the culture is very young. After resuscitation split cells 1:3 to 1:4 and culture for at least one week with 1-2 media changes before shipping.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service