Skip to Content
Merck
All Photos(1)

Key Documents

BCR723

Road dust (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental

format

neat

Analysis Note

For more information please see:
BCR723

Citation

Platinum-group element concentrations in BCR-723

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross A Sutherland
Analytica chimica acta, 582(2), 201-207 (2007-03-28)
BCR-723 is the only environmental certified reference material for platinum-group elements (PGEs) Pt, Pd and Rh. It has been an integral component of quality control/quality assurance procedures in environmental laboratories measuring PGEs, since 2001. Here, we present an extensive quantitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service