16336
Nutrient Broth No 3, Vegitone
suitable for microbiology, NutriSelect® Plus
Synonym(s):
Vegitone Nutrient Broth No 3
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
peptone (vegetable), 5.0 g/L
sodium chloride, 5.0 g/L
vegetable extract, 1.5 g/L
yeast extract, 1.5 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.4±0.2 (25 °C)
application(s)
agriculture
clinical testing
environmental
food and beverages
life science and biopharma
pharmaceutical
water monitoring
microbiology
suitability
nonselective for bacteria (General Media)
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service