Skip to Content
Merck
All Photos(2)

Key Documents

371724

Sigma-Aldrich

GPR119 Agonist, AS1269574

The GPR119 Agonist, AS1269574, also referenced under CAS 330981-72-1, controls the biological activity of GPR119. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

GPR119 Agonist, AS1269574, 2-((2-(4-Bromophenyl)-6-methylpyrimidin-4-yl)amino)ethanol, G-Protein Coupled Receptor 119 Agonist, GPCR2 Agonist, MGC119957 Agonist, SNORF25 Agonist, GPCR2 Agonist, MGC119957 Agonist, SNORF25 Agonist, 2-((2-(4-Bromophenyl)-6-methylpyrimidin-4-yl)amino)ethanol, G-Protein Coupled Receptor 119 Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14BrN3O
CAS Number:
Molecular Weight:
308.17
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.51

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

amber

solubility

DMSO: >10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

BrC(C=C1)=CC=C1C2=NC(C)=CC(NCCO)=N2

InChI

1S/C13H14BrN3O/c1-9-8-12(15-6-7-18)17-13(16-9)10-2-4-11(14)5-3-10/h2-5,8,18H,6-7H2,1H3,(H,15,16,17)

InChI key

DUKPGOOUJNUIOI-UHFFFAOYSA-N

General description

A pyrimidine compound that acts as an effective GPR119 agonist (EC50 = 2.5 µM in stimulating cAMP in hGPR119-expressing HEK293 cells) and reportedly exhibits no activity toward β-adrenergic or GLP-1 receptors. Shown to enhance 16.8 mM glucose-induced insulin secretion from murine MIN-6 β-cells (mU/20 min = 0.22, 0.29, 0.44, respectively, with 0, 1, 10 µM AS1269574) in vitro and improve glucose tolerance in normal mice (26% decrease and 60% increase, respectively, of control blood glucose and insulin AUC0-1h in OGTT; 100 mg/kg; p.o.) without affecting basal blood glucose level in either fed or fasting mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Yoshida, S., et cl. 2010. Biochem. Biophys. Res. Commun.400, 437.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service