Skip to Content
Merck
All Photos(1)

Key Documents

03-185

Sigma-Aldrich

Histone H2B human recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160405
NACRES:
NA.84
technique(s):
activity assay: suitable
western blot: suitable
application:
WB
activity assay
citations:

antibody form

purified antibody

Quality Level

Assay

>50% (SDS-PAGE)

technique(s)

activity assay: suitable
western blot: suitable

General description

Histone H2B is one of four core histones that form an octameric structure required for the packaging of DNA into nucleosomes, the most basic level of chromatin arrangement. Histones undergo a number of post-translational modifications (PTM) at their N- and C-terminal tails which can influence chromatin structure and stability and consequently, transcription, and DNA replication and repair. The most widely studied of these modifications are methylation, phosphorylation, and acetylation which may produce different effects on gene regulation depending on the number and pattern of modifications. Histone H2B undergoes acetylation at lysines 5, 15, and 20. It is also phosphorylated in apoptotic cells during DNA fragmentation, possibly in a caspase-dependent manner.
Human recombinant H2B expressed in E. Coli

Application

Chromatin assembly, Western blot (control), Kinase Assay

Physical properties

~17 kDa

Physical form

Liquid in Dulbecco′s PBS. Frozen solution.

Storage and Stability

Stable for 12 months at -80°C from date of receipt. Avoid multiple freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service