Merck
All Photos(5)
Key Documents
BAF378740001
SP Bel-Art® Sterileware Powder Thief
capacity × L 1 mL (0.03 oz) × 19.75 in. (50 cm), pack of 20 ea (individually wrapped)
Synonym(s):
sampler
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 20 ea (individually wrapped)
manufacturer/tradename
Bel-Art F37874-0001
capacity × L
1 mL (0.03 oz) × 19.75 in. (50 cm)
General description
Available in two lengths each with four tip volumes, these sterile powder thieves easily sample from targeted locations within bulk powders. Used by pharmaceutical, food, cosmetic and chemical companies.
Features and Benefits
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)