Skip to Content
Merck
All Photos(1)

Documents

694622

Sigma-Aldrich

Lumidot CdSe/ZnS

590, core-shell type quantum dots, 5 mg/mL in toluene

Synonym(s):

QD, Quantum dot semiconductor nanocrystal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

Quality Level

form

dispersion

availability

not available in USA

concentration

5 mg/mL in toluene

particle size

4.0 nm

bp

110 °C

density

0.865 g/mL at 25 °C

ε (extinction coefficient)

1.60 x 105 M-1cm-1

fluorescence

λex 575 nm; λem 590 nm FWHM <40 nm, quantum yield ≥30%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Orange emitting quantum dot nanoparticles

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3

Flash Point(F)

closed cup

Flash Point(C)

closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service