Skip to Content
Merck
All Photos(1)

Key Documents

604151

Sigma-Aldrich

Phthaloyl chloride-2,2′-13C2

99 atom % 13C

Synonym(s):

1,2-Benzenedi(carbonyl-13C) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C6Cl2O2
CAS Number:
Molecular Weight:
200.98
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

bp

269-271 °C (lit.)

mp

6-12 °C (lit.)

density

1.423 g/mL at 25 °C

mass shift

M+2

SMILES string

Cl[13C](=O)c1ccccc1[13C](Cl)=O

InChI

1S/C8H4Cl2O2/c9-7(11)5-3-1-2-4-6(5)8(10)12/h1-4H/i7+1,8+1

InChI key

FYXKZNLBZKRYSS-BFGUONQLSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service