Skip to Content
Merck
All Photos(2)

Key Documents

233536

Sigma-Aldrich

4-Decylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9C6H4NH2
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

218-220 °C/14 mmHg (lit.)

mp

25 °C (lit.)

SMILES string

CCCCCCCCCCc1ccc(N)cc1

InChI

1S/C16H27N/c1-2-3-4-5-6-7-8-9-10-15-11-13-16(17)14-12-15/h11-14H,2-10,17H2,1H3

InChI key

WGENWPANMZLPIH-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speciation of nitrogen compounds in gasoline and diesel range process streams by capillary column gas chromatography with chemiluminescence detection.
ChawlaB.
Journal of Chromatographic Science, 35(3), 97-104 (1997)
New chromoreactands for the detection of aldehydes, amines and alcohols.
Mohr GJ.
Sensors and Actuators B, Chemical, 90(1), 31-36 (2003)
Plasticization of a poly (ethyl acrylate) ionomer by an alkyl aniline.
Tong X and Bazuin CG.
Journal of Polymer Science. Part B, Polymer Physics, 30(4), 389-399 (1992)
T Flinois et al.
Colloids and surfaces. B, Biointerfaces, 190, 110924-110924 (2020-03-09)
Quartz crystal microbalance studies have been carried out to monitor the fusion of lipid vesicles (pure 1,2-dimyristoyl-sn-glycero-3-phosphocholine, DMPC) and mixed vesicles (DMPC and 4-decylaniline). In order to increase the stability of the lipid deposits onto the electrodes, we have developed
Kyle A Brown et al.
Nature methods, 16(5), 417-420 (2019-04-17)
We report the identification of a photocleavable anionic surfactant, 4-hexylphenylazosulfonate (Azo), which can be rapidly degraded by ultraviolet irradiation, for top-down proteomics. Azo can effectively solubilize proteins with performance comparable to that of sodium dodecyl sulfate (SDS) and is compatible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service