Skip to Content
Merck
All Photos(1)

Key Documents

EP5702713007

Eppendorf® Adapter for 90 mL rectangular bucket for use with standard tubes

Holds 20 mL tubes, pack of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

feature

Holds 20 mL tubes

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5702713007

technique(s)

centrifugation: suitable

compatibility

for use with 90 mL rectangular buckets

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service