EP0030108434
Eppendorf® Protein LoBind Tubes
PCR clean, withstand 18,000 x g, 0.5 mL, pack of 500 ea
Synonym(s):
Eppendorf® Protein LoBind tubes
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Recommended Products
material
cap (push fit)
colorless
conical bottom
description
Low retention
sterility
non-sterile
feature
DNase free
PCR clean
packaging
pack of 500 ea
manufacturer/tradename
Eppendorf® 0030108434
capacity
0.5 mL
suitability
suitable for PCR
General description
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Features and Benefits
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)