MABN889-AF488
Anti-C9ORF72/C9RANT (poly-GA) Antibody, clone 5E9 Antibody, Alexa Fluor™ 488 Conjugate
clone 5E9, from mouse, ALEXA FLUOR™ 488
Synonym(s):
C9ORF72/C9RANT (poly-GA), Protein C9orf72
About This Item
biological source
mouse
Quality Level
conjugate
ALEXA FLUOR™ 488
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
5E9, monoclonal
species reactivity
human
species reactivity (predicted by homology)
all (based on 100% sequence homology)
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... C9ORF72(203228)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected C9ORF72/C9RANT (poly-GA) Western Blotting applications (Mackenzie, I.R., et. al. (2013). Acta Neuropathol. 126(6):859-79).
Neuroscience
Quality
Western Blotting Analysis: 4 µg/mL of this antibody detected Gly-Ala repeats in GST-fusion recombinant C9orf72 protein.
Target description
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service