Z740952
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 50 mL, O.D. × L 25 mm × 150 mm, round bottom
All Photos(1)
About This Item
UNSPSC Code:
41121706
NACRES:
NB.11
Recommended Products
material
borosilicate glass tube
round bottom
screw top cap
screw top neck
feature
neck thread external thread
packaging
pack of 48 ea
manufacturer/tradename
Sigma-Aldrich
O.D. × L
25 mm × 150 mm
capacity
50 mL
Related Categories
General description
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)