854188
Crimp seals, 11 mm, with PTFE/rubber septa
gold aluminum seal, size × thickness 11 mm × 1.0 mm, septum (TFE/butyl/red rubber), pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
gold aluminum seal
septum (TFE/butyl/red rubber)
feature
closure type crimp seal
hardness
45 (Durometer: shore A)
packaging
pkg of 100 ea
size × thickness
11 mm × 1.0 mm
fitting
thread for 11 mm
Looking for similar products? Visit Product Comparison Guide
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service