814907
TOYOPEARL® AF-Chelate-650M Bulk Media
phase IDA, iminodiacetic acid, bottle of 1 L, 65 μm particle size
Synonym(s):
TOYOPEARL® AF-Chelate-650M Bulk Media
About This Item
description
Clean In Place (CIP) (0.5 M NaOH or 0.1 M HCl)
Shipping solvent (20% (v/v) ethanol)
phase filter (Chelate)
product line
TOYOPEARL®
form
slurry
packaging
bottle of 1 L
manufacturer/tradename
Tosoh 14907
extent of labeling
25-45 μeq/mL ligand density
parameter
pressure (max. 3 bar)
technique(s)
LPLC: suitable
matrix
hydroxylated methacrylic polymer particle platform
matrix active group
IDA phase, iminodiacetic acid
particle size
65 μm
pore size
1000 Å
pH
2-12
separation technique
affinity
Looking for similar products? Visit Product Comparison Guide
General description
Specifications
Physical form
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service