Skip to Content
Merck
All Photos(1)

Key Documents

55069

Tubing Adapter

M6 female, 10-32 male

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

PEEK adapter (one-piece design)

description

for chromatography applications

packaging

pkg of 1 ea

parameter

1000 psi max. pressure (69 bar)

adapter swept volume

6.70 μL

bore diam.

0.030 in. (0.75 mm)

fitting

10-32 male
M6 female

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service