U6758
UCL 2077
≥98% (HPLC), solid
Synonym(s):
(3-Triphenylmethylaminomethyl)pyridine
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
solid
color
white
solubility
DMSO: soluble >20 mg/mL
H2O: insoluble
SMILES string
C(NC(c1ccccc1)(c2ccccc2)c3ccccc3)c4cccnc4
InChI
1S/C25H22N2/c1-4-12-22(13-5-1)25(23-14-6-2-7-15-23,24-16-8-3-9-17-24)27-20-21-11-10-18-26-19-21/h1-19,27H,20H2
InChI key
PQFNWDHABGBCHB-UHFFFAOYSA-N
Application
Biochem/physiol Actions
Preparation Note
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service