Skip to Content
Merck
All Photos(1)

Key Documents

SAB4701082

Sigma-Aldrich

Monoclonal Anti-CD253-APC antibody produced in mouse

clone 2E5

Synonym(s):

Apo-2 ligand, Apo-2L, Protein TRAIL, TNF-related apoptosis-inducing ligand, Tumor necrosis factor ligand superfamily member 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E5, monoclonal

form

buffered aqueous solution

concentration

0.1 mg/mL

technique(s)

flow cytometry: 4 μg/mL

accession no.

BC020220

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TNFSF10(8743)

Related Categories

General description

The antibody 2E5 reacts with TRAIL (APO-2L), a 21 kDa cytotoxic protein, activator of rapid apoptosis in tumor cells. TRAIL is mainly expressed in spleen, lung, prostate and also in many other tissues.

Immunogen

Recombinant soluble fragment (aa 95-281) of human TRAIL.

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service