Skip to Content
Merck
All Photos(1)

Key Documents

KHOS-240S

86112810, human bone, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

KHOS-240S, 86112810

biological source

human bone

growth mode

Adherent

karyotype

2n = 46, aneuploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian osteosarcoma

Cell Line Description

A revertant line of the KHOS/NP cell line (Sigma Catalogue number. 84102903). Established after overnight incubation at 40.5°C and then sub-cloned at 36°C. The line has similar properties to the parent HOS cell line. Cells are non-tumourigenic in nude mice and no longer possess a rescuable MSV genome.

Application

Tumourigenicity, transformation, biochemical and virus studies: MSV

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 10,13
D5S818: 13
D7S820: 11,12
THO1: 6
TPOX: 11
vWA: 18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service