Skip to Content
Merck
All Photos(1)

Key Documents

90119

Sigma-Aldrich

N-Methyl-L-valine

≥98% (TLC)

Synonym(s):

(S)-3-Methyl-2-(methylamino)butyric acid, N-Me-Val-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721712
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (TLC)

form

powder with small lumps

optical activity

[α]/D +25.0±1.0°, c = 1 in 1 M HCl

composition

carbon, 54.1-55.7%
nitrogen, 10.4-11.0%

technique(s)

ligand binding assay: suitable

color

white

SMILES string

CN[C@@H](C(C)C)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)5(7-3)6(8)9/h4-5,7H,1-3H3,(H,8,9)/t5-/m0/s1

InChI key

AKCRVYNORCOYQT-YFKPBYRVSA-N

Biochem/physiol Actions

N-Methyl amino acids play an important role in investigations on prevention or therapy of Alzheimer′s disease, neurofibrillary tangles or amyloid plaque are physiological characteristics of this disease and ?-amyloid(40) fibrillogenesis and disassembly of ?-amyloid(40) fibrils is inhibited by short ?-amyloid congeners containing N-methyl amino acids at alternate residue.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franco Cataldo et al.
Life (Basel, Switzerland), 3(3), 449-473 (2013-01-01)
A series of non-proteinogenic amino acids, most of them found quite commonly in the meteorites known as carbonaceous chondrites, were subjected to solid state radiolysis in vacuum to a total radiation dose of 3.2 MGy corresponding to 23% of the
David R Calabrese et al.
Nature communications, 9(1), 4229-4229 (2018-10-14)
G-quadruplexes (G4s) are noncanonical DNA structures that frequently occur in the promoter regions of oncogenes, such as MYC, and regulate gene expression. Although G4s are attractive therapeutic targets, ligands capable of discriminating between different G4 structures are rare. Here, we
D J Gordon et al.
Biochemistry, 40(28), 8237-8245 (2001-07-11)
A potential goal in the prevention or therapy of Alzheimer's disease is to decrease or eliminate neuritic plaques composed of fibrillar beta-amyloid (Abeta). In this paper we describe N-methyl amino acid containing congeners of the hydrophobic "core domain" of Abeta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service