Skip to Content
Merck
All Photos(1)

Key Documents

61726

Sigma-Aldrich

N-Lauroyl-L-alanine

≥99.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CONHCH(CH3)COOH
CAS Number:
Molecular Weight:
271.40
Beilstein:
1728158
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

description

non-ionic

Assay

≥99.0% (TLC)

form

powder

mol wt

271.40 g/mol

technique(s)

protein quantification: suitable

color

white

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(=O)N[C@@H](C)C(O)=O

InChI

1S/C15H29NO3/c1-3-4-5-6-7-8-9-10-11-12-14(17)16-13(2)15(18)19/h13H,3-12H2,1-2H3,(H,16,17)(H,18,19)/t13-/m0/s1

InChI key

UYTOHYBIBPDOKX-ZDUSSCGKSA-N

Biochem/physiol Actions

N-Lauroyl-L-alanine is a fatty acid amide that may be used to support the physical gelation of binary mixtures of aliphatic and aromatic hydrocarbon solvents with potential use in injectable biodegradable drug delivery systems.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keke Wang et al.
International journal of pharmaceutics, 404(1-2), 176-179 (2010-11-03)
The purpose of the present study was to develop a novel, simple and determination-of-electrical-conductivity-based method to trace the gelling process of injectable biodegradable in situ forming organogels after administration. The electrical conductivity of pH 7.4 PBS solution with different amount
Michael Lau et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 138, 105038-105038 (2019-08-10)
Antioxidants play a vital role in scavenging reactive oxygen species (ROS) produced by the reduction of molecular oxygen from various cellular mechanisms. Under oxidative stress, an increase in the levels of ROS overwhelms the antioxidant response, causing oxidative damage to
Physical gelation of binary mixtures of hydrocarbons mediated by n-lauroyl-L-alanine and characterization of their thermal and mechanical properties.
Bhattacharya S, Pal A.
The Journal of Physical Chemistry B, 112, 491804927-491804927 (2998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service