428361
2,7,12,17-Tetra-tert-butyl-5,10,15,20-tetraaza-21H,23H-porphine
Dye content 85 %
About This Item
Recommended Products
Assay
≥83.0% (HPLC)
Quality Level
form
powder or crystals
composition
Dye content, 85%
mp
182 °C (dec.) (lit.)
λmax
618.0-624.0 nm in hexane
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
CC(C)(C)c1cc2nc1nc3cc(c(nc4cc(c(n4)nc5cc(c(n2)[nH]5)C(C)(C)C)C(C)(C)C)[nH]3)C(C)(C)C
InChI
1S/C32H42N8/c1-29(2,3)17-13-21-33-25(17)37-22-14-18(30(4,5)6)27(34-22)39-24-16-20(32(10,11)12)28(36-24)40-23-15-19(31(7,8)9)26(35-23)38-21/h13-16H,1-12H3,(H2,33,34,35,36,37,38,39,40)
InChI key
DKCJDGYFHWIGBO-UHFFFAOYSA-N
Related Categories
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service