Skip to Content
Merck
All Photos(1)

Key Documents

XX2504754

Millipore

Funnel Cover, 47mm, silicone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

clear device

sterility

non-sterile

input

sample type bottled water
sample type beer
sample type pharmaceutical(s)
sample type cosmetics

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

shipped in

ambient

General description

For general filtration applications and filtration of samples for particulate or biological contamination analysis

Application

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing , Water Monitoring

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service