8.55085
HMPA-NovaGel
Novabiochem®
Synonym(s):
4-Hydroxymethylphenoxyacetyl NovaGel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111023
NACRES:
NA.22
Quality Level
product line
NovaGel
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
alcohol
storage temp.
2-8°C
Related Categories
General description
This resin is prepared by derivatization of aminomethyl NovaGel with the TFA-labile 4-hydroxymethylphenoxyacetic acid linker. NovaGel resins combine the high functionality of polystyrene resins with the excellent swelling properties of PEG-PS type supports [1].
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Literature references
[1] J. H. Adams, et al. (1998) J. Org. Chem., 63, 3706.
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Literature references
[1] J. H. Adams, et al. (1998) J. Org. Chem., 63, 3706.
Linkage
Replaces: 01-64-0284
Analysis Note
Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.50 - 0.80 mmol/g
Swelling Volume (in DMF): lot specific result
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.50 - 0.80 mmol/g
Swelling Volume (in DMF): lot specific result
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service