Skip to Content
Merck
All Photos(1)

Key Documents

8.18773

Sigma-Aldrich

4-Nonanol

for synthesis

Synonym(s):

4-Nonanol, Pentyl propyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH2CH2CH3
CAS Number:
Molecular Weight:
144.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
227-679-8
NACRES:
NA.22

Quality Level

Assay

≥96.5% (GC)

form

liquid

transition temp

flash point 74 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CCCCC)CCC

InChI

1S/C9H20O/c1-3-5-6-8-9(10)7-4-2/h9-10H,3-8H2,1-2H3

InChI key

IXUOEGRSQCCEHB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.821 - 0.823
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Coelho et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-07)
Knowledge of composition of beverages volatile fraction is essential for understanding their sensory attributes. Analysis of volatile compounds predominantly resorts to gas chromatography coupled with mass spectrometry (GC-MS). Often a previous concentration step is required to quantify compounds found at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service