Skip to Content
Merck
All Photos(1)

Key Documents

8.14501

Sigma-Aldrich

2-Chloroethanesulfonic acid sodium salt monohydrate

for synthesis

Synonym(s):

2-Chloroethanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4ClNaO3S · xH2O
CAS Number:
Molecular Weight:
166.56 (anhydrous basis)
UNSPSC Code:
12352107
EC Index Number:
239-508-4
NACRES:
NA.22

Quality Level

solubility

100 g/L

storage temp.

2-30°C

Analysis Note

Assay (acidimetric, calc. on anhydrous substance): ≥ 98.0 %
Water (K. F.): 9.0 - 11.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service