Skip to Content
Merck
All Photos(1)

Key Documents

8.14138

Sigma-Aldrich

3-Nitropropanol

for synthesis

Synonym(s):

3-Nitropropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
MDL number:
UNSPSC Code:
12352104
EC Index Number:
246-716-9
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

potency

77 mg/kg LD50, oral (Rat)

bp

138-140 °C/43 hPa

density

1.21 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])CCCO

InChI

1S/C3H7NO3/c5-3-1-2-4(6)7/h5H,1-3H2

InChI key

YISPKQZWSIMXMX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.208 - 1.212
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service