Skip to Content
Merck
All Photos(1)

Key Documents

878120P

Avanti

C18 Lyso PAF

Avanti Research - A Croda Brand 878120P, powder

Synonym(s):

1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H56NO6P
CAS Number:
Molecular Weight:
509.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 5 mg (878120P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878120P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCCCCCCCCCCCC

InChI

1S/C26H56NO6P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22-31-24-26(28)25-33-34(29,30)32-23-21-27(2,3)4/h26,28H,5-25H2,1-4H3/t26-/m1/s1

InChI key

XKBJVQHMEXMFDZ-AREMUKBSSA-N

General description

C18-lysoPAF, also known as 1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphocholine, is a synthetic lyso-platelet activating factor (PAF). It is a precursor and metabolite of 1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine (C18:0 PAF). Lyso-platelet activating factor (PAF) is synthesized from PAF acetylhydrolases by the degradation of PAF. Additionally, it is also synthesized in the remodeling pathway from 1-O-alkyl-2-arachidonoyl-sn-glycero-3-phosphocholine (1-alkyl-phosphatidylcholine) by the enzyme phospholipase.[1]

Application

C18-lysoPAF (1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphocholine) has been used to study its effect on human eosinophil degranulation.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (878120P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideo Shindou et al.
The Journal of biological chemistry, 282(9), 6532-6539 (2006-12-22)
Platelet-activating factor (PAF) is a potent proinflammatory lipid mediator eliciting a variety of cellular functions. Lipid mediators, including PAF are produced from membrane phospholipids by enzymatic cascades. Although a G protein-coupled PAF receptor and degradation enzymes have been cloned and
Kimberly D Dyer et al.
Journal of immunology (Baltimore, Md. : 1950), 184(11), 6327-6334 (2010-04-28)
Platelet-activating factor (PAF [1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine]) is a phospholipid mediator released from activated macrophages, mast cells, and basophils that promotes pathophysiologic inflammation. Eosinophil responses to PAF are complex and incompletely elucidated. We show in this article that PAF and its 2-deacetylated metabolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service