Key Documents
Z260517
Sigma-Aldrich
sidEntry glove box
a clear acrylic glove box which features a large side entry, gasketed door, and a shelf rack
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H50026-0000
L × W × H
30 in. × 24 in. × 24 in.
General description
The Scienceware sidEntry glove box is a clear acrylic glove box which features a large side entry, gasketed door, and a shelf rack to store small items. Glove box stability is assured by four adjustable levelers that also level the box and all its contents. The sidEntry glove box provides an isolated work environment for processing samples or handling air-sensitive materials while maintaining an anaerobic or other gas specific environment within the glove box.
  • 8 in. glove ports accept standard dry-box gloves without clamps or O-rings
  • Small enough to move from area to area
  • Side door: 17 1/2 in. × 17 1/2 in.
Legal Information
sidEntry is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)