Key Documents
Z136085
Bel-Bulb pipettor
vinyl bulb, fits 6-8 mm pipettes, L × diam. 70 mm × 5 mm
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
vinyl bulb
feature
fits 6-8 mm pipettes
packaging
pack of 2 ea
manufacturer/tradename
Bel-Art F37887-0000
L × diam.
70 mm × 5 mm
Related Categories
General description
Flexible vinyl bulb. A soft tapered chuck allows bulb to be held against pipette for suctioning.
Legal Information
Bel-Bulb is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)