All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H3BrF2O2
CAS Number:
Molecular Weight:
237.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
118-122 °C (lit.)
SMILES string
OC(=O)c1cc(F)c(F)cc1Br
InChI
1S/C7H3BrF2O2/c8-4-2-6(10)5(9)1-3(4)7(11)12/h1-2H,(H,11,12)
InChI key
DGCBGBZYTNTZJH-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service