Skip to Content
Merck
All Photos(1)

Key Documents

510149

Sigma-Aldrich

tert-Butyl indoline-1-carboxylate

98%

Synonym(s):

N-(tert-Butoxycarbonyl)-2,3-dihydroindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO2
CAS Number:
Molecular Weight:
219.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

bp

83-84 °C/0.1 mmHg (lit.)

mp

46-50 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCc2ccccc12

InChI

1S/C13H17NO2/c1-13(2,3)16-12(15)14-9-8-10-6-4-5-7-11(10)14/h4-7H,8-9H2,1-3H3

InChI key

GWAXLDLPPZPQLO-UHFFFAOYSA-N

General description

tert -Butyl indoline-1-carboxylate is an N-substituted indoline derivative.

Application

  • Reactant in preparation of aryl alkyl amines via alkylation reactions catalyzed by Pd
  • Reactant in preparation of allyl- and arylindolines
  • Reactant in modular indole synthesis of the highly strained CDEF parent tetracycle of nodulisporic acids A and B
  • Reactant in asymmetric synthesis of β-amino esters via rhodium prolinate complex-catalyzed α-C-H activation / carbenoid insertion reactions
  • Reactant in preparation of substituted indolines and tetrahydroquinolines via cycloaddition approach

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service